Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL2L13
Entrez Gene ID
23786
Gene Name
BCL2 like 13
Gene Aliases
BCL-RAMBO, Bcl2-L-13, MIL1
Chromosome Location
Chr.22: 17628855 - 17730855 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270726.1 NP_001257655.1 4 - 5 642 120
NM_001270729.1 NP_001257658.1 4 - 5 656 120
NM_001270730.1 NP_001257659.1 3 - 4 485 120
NM_015367.3 NP_056182.2 5 - 6 813 120
XM_011546119.1 XP_011544421.1 5 - 6 1176 120
XM_011546120.1 XP_011544422.1 5 - 6 1178 120
XM_011546121.2 XP_011544423.1 6 - 7 1161 120
XM_017028726.1 XP_016884215.1 7 - 8 1462 120
XM_017028728.1 XP_016884217.1 5 - 6 1190 120
XM_017028731.1 XP_016884220.1 4 - 5 1019 120
XM_017028733.1 XP_016884222.1 5 - 6 813 120
GenBank mRNA AF246665.1 - 6 - 7 1199 120
AF325209.1 - 4 - 5 460 120
AK056248.1 - 4 - 5 499 120
AK314489.1 - 5 - 6 640 120
BC007658.2 - 5 - 6 572 120
BX354578.2 - 4 - 5 521 120
BX407004.2 - 4 - 5 395 120
CR456521.1 - 5 - 6 716 120
DA154984.1 - 3 - 4 312 120

View Full Product Details