The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001314030.1 | NP_001300959.1 | 6 - 7 | 1238 | 62 |
NM_012284.2 | NP_036416.1 | 6 - 7 | 1242 | 62 | |
XM_011538085.2 | XP_011536387.1 | 6 - 7 | 1858 | 62 | |
XM_011538086.2 | XP_011536388.1 | 6 - 7 | 1854 | 62 | |
XM_017019096.1 | XP_016874585.1 | 5 - 6 | 1624 | 62 | |
XM_017019097.1 | XP_016874586.1 | 6 - 7 | 862 | 62 | |
GenBank mRNA | AB022696.1 | - | 6 - 7 | 1017 | 62 |
AB033108.1 | - | 6 - 7 | 1242 | 62 | |
BC150602.1 | - | 6 - 7 | 1123 | 62 |