Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERCC1
Entrez Gene ID
2067
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Gene Aliases
COFS4, RAD10, UV20
Chromosome Location
Chr.19: 45407333 - 45478866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166049.1 NP_001159521.1 1 - 2 140 59
NM_001983.3 NP_001974.1 1 - 2 140 59
XM_005258634.1 XP_005258691.1 1 - 2 71 59
XM_005258637.1 XP_005258694.1 1 - 2 71 59
GenBank mRNA AB069681.1 - 1 - 2 60 59
AF001925.1 - 1 - 2 136 59
AF433652.1 - 1 - 2 115 59
AK314884.1 - 1 - 2 72 59
BC008930.2 - 1 - 2 56 59
BQ671113.1 - 1 - 2 140 59
M13194.1 - 1 - 2 136 59
M28650.1 - 1 - 2 136 59

View Full Product Details