Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC5
Entrez Gene ID
56915
Gene Name
exosome component 5
Gene Aliases
RRP41B, RRP46, Rrp46p, hRrp46p, p12B
Chromosome Location
Chr.19: 41386370 - 41397351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020158.3 NP_064543.3 3 - 4 402 59
GenBank mRNA AF281134.1 - 3 - 4 402 59
BC007742.2 - 3 - 4 385 59
BC107696.1 - 3 - 4 407 59

View Full Product Details