The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015139.2 | NP_055954.1 | 7 - 8 | 1017 | 92 |
XM_006710478.1 | XP_006710541.1 | 7 - 8 | 1017 | 92 | |
XM_011541070.1 | XP_011539372.1 | 7 - 8 | 1017 | 92 | |
GenBank mRNA | AB044343.1 | - | 7 - 8 | 665 | 92 |
AK289800.1 | - | 7 - 8 | 1015 | 92 | |
AK296449.1 | - | 6 - 7 | 488 | 92 | |
BC093786.1 | - | 7 - 8 | 659 | 92 | |
BC112031.1 | - | 7 - 8 | 671 | 92 | |
D87449.1 | - | 7 - 8 | 719 | 92 |