Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSSCA1
Entrez Gene ID
10534
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
Gene Aliases
p27
Chromosome Location
Chr.11: 65570459 - 65571888 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006396.2 NP_006387.1 1 - 2 54 61
GenBank mRNA AB001740.1 - 1 - 2 41 61
BC014791.2 - 1 - 2 40 61
BP351453.1 - 1 - 2 55 61

View Full Product Details