Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNK1
Entrez Gene ID
65125
Gene Name
WNK lysine deficient protein kinase 1
Gene Aliases
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65
Chromosome Location
Chr.12: 752552 - 911452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184985.1 NP_001171914.1 26 - 27 8067 66
NM_014823.2 NP_055638.2 24 - 25 6543 66
NM_018979.3 NP_061852.3 26 - 27 7287 66
NM_213655.4 NP_998820.3 26 - 27 8043 66
XM_006719003.2 XP_006719066.1 26 - 27 7609 66
XM_011520997.2 XP_011519299.1 27 - 28 8897 66
XM_011520998.2 XP_011519300.1 27 - 28 8848 66
XM_011520999.2 XP_011519301.1 27 - 28 8848 66
XM_011521001.2 XP_011519303.1 26 - 27 8572 66
XM_011521002.2 XP_011519304.1 26 - 27 8389 66
XM_011521003.2 XP_011519305.1 25 - 26 8113 66
XM_011521004.2 XP_011519306.1 25 - 26 8110 66
XM_011521005.2 XP_011519307.1 24 - 25 7335 66
XM_011521006.2 XP_011519308.1 25 - 26 7528 66
XM_011521007.2 XP_011519309.1 25 - 26 7525 66
XM_011521008.2 XP_011519310.1 23 - 24 6790 66
XM_011521009.2 XP_011519311.1 23 - 24 6787 66
XM_017019834.1 XP_016875323.1 25 - 26 7150 66
XM_017019835.1 XP_016875324.1 24 - 25 7069 66
XM_017019836.1 XP_016875325.1 24 - 25 7066 66
XM_017019837.1 XP_016875326.1 24 - 25 6874 66
XM_017019838.1 XP_016875327.1 24 - 25 6871 66
GenBank mRNA AB002342.2 - 24 - 25 5698 66
AJ296290.1 - 26 - 27 6644 66
BC141881.1 - 18 - 19 4111 66

View Full Product Details