Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNK1
Entrez Gene ID
65125
Gene Name
WNK lysine deficient protein kinase 1
Gene Aliases
HSAN2, HSN2, KDP, PPP1R167, PRKWNK1, PSK, p65
Chromosome Location
Chr.12: 752552 - 911452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184985.1 NP_001171914.1 3 - 4 1788 114
NM_014823.2 NP_055638.2 3 - 4 1788 114
NM_018979.3 NP_061852.3 3 - 4 1788 114
NM_213655.4 NP_998820.3 3 - 4 1788 114
XM_006719003.2 XP_006719066.1 3 - 4 2113 114
XM_011520997.2 XP_011519299.1 3 - 4 2159 114
XM_011520998.2 XP_011519300.1 3 - 4 2113 114
XM_011520999.2 XP_011519301.1 3 - 4 2113 114
XM_011521000.2 XP_011519302.1 3 - 4 2113 114
XM_011521001.2 XP_011519303.1 3 - 4 2113 114
XM_011521002.2 XP_011519304.1 3 - 4 2113 114
XM_011521003.2 XP_011519305.1 3 - 4 2113 114
XM_011521004.2 XP_011519306.1 3 - 4 2113 114
XM_011521006.2 XP_011519308.1 3 - 4 2113 114
XM_011521007.2 XP_011519309.1 3 - 4 2113 114
XM_011521008.2 XP_011519310.1 3 - 4 2113 114
XM_011521009.2 XP_011519311.1 3 - 4 2113 114
XM_017019834.1 XP_016875323.1 3 - 4 2113 114
XM_017019835.1 XP_016875324.1 3 - 4 2113 114
XM_017019836.1 XP_016875325.1 3 - 4 2113 114
XM_017019837.1 XP_016875326.1 3 - 4 2113 114
XM_017019838.1 XP_016875327.1 3 - 4 2113 114
XM_017019839.1 XP_016875328.1 3 - 4 2113 114
GenBank mRNA AB002342.2 - 3 - 4 943 114
AF061944.1 - 3 - 4 1788 114
AJ296290.1 - 3 - 4 1145 114
CD629562.1 - 1 - 2 112 114
JQ358908.1 - 3 - 4 1145 114

View Full Product Details