The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316341.1 | NP_001303270.1 | 10 - 11 | 1300 | 85 |
NM_002816.4 | NP_002807.1 | 8 - 9 | 1016 | 85 | |
NM_174871.3 | NP_777360.1 | 7 - 8 | 956 | 85 | |
GenBank mRNA | AB003103.1 | - | 8 - 9 | 951 | 85 |
AK091198.1 | - | 7 - 8 | 934 | 85 | |
AK307821.1 | - | 8 - 9 | 966 | 85 | |
BC019062.2 | - | 8 - 9 | 947 | 85 | |
BC065826.1 | - | 10 - 11 | 1220 | 85 | |
KU178288.1 | - | 8 - 9 | 908 | 85 |