The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257378.1 | NP_001244307.1 | 7 - 8 | 727 | 85 |
NM_001257379.1 | NP_001244308.1 | 6 - 7 | 557 | 85 | |
NM_001257382.1 | NP_001244311.1 | 6 - 7 | 627 | 85 | |
NM_174975.4 | NP_777635.1 | 5 - 6 | 509 | 85 | |
XM_011530128.2 | XP_011528430.1 | 5 - 6 | 477 | 85 | |
GenBank mRNA | AK131358.1 | - | 6 - 7 | 531 | 85 |
AY158086.1 | - | 5 - 6 | 420 | 85 | |
AY240872.1 | - | 5 - 6 | 509 | 85 | |
BC069641.1 | - | 5 - 6 | 432 | 85 | |
BC101001.2 | - | 7 - 8 | 666 | 85 | |
BC101002.1 | - | 6 - 7 | 496 | 85 | |
BC101003.1 | - | 6 - 7 | 566 | 85 | |
BC101004.1 | - | 5 - 6 | 448 | 85 |