The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015918.3 | NP_057002.2 | 3 - 4 | 343 | 96 |
GenBank mRNA | AF070660.1 | - | 3 - 4 | 327 | 96 |
AF117232.1 | - | 3 - 4 | 327 | 96 | |
AJ306296.1 | - | 3 - 4 | 330 | 96 | |
BC012505.1 | - | 3 - 4 | 330 | 96 | |
BI668578.1 | - | 3 - 4 | 350 | 96 | |
CB995722.1 | - | 3 - 4 | 465 | 96 |