The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278182.1 | NP_001265111.1 | 3 - 4 | 1578 | 109 |
NM_001278183.1 | NP_001265112.1 | 3 - 4 | 562 | 109 | |
NM_005442.3 | NP_005433.2 | 3 - 4 | 1578 | 109 | |
XM_005265510.3 | XP_005265567.1 | 3 - 4 | 1576 | 109 | |
GenBank mRNA | AB031038.1 | - | 3 - 4 | 1157 | 109 |
AK297389.1 | - | 4 - 5 | 578 | 109 | |
AK299499.1 | - | 4 - 5 | 960 | 109 | |
AK299695.1 | - | 4 - 5 | 644 | 109 | |
AK316166.1 | - | 3 - 4 | 562 | 109 | |
BC025363.1 | - | 2 - 3 | 271 | 109 | |
BC037568.1 | - | 3 - 4 | 1181 | 109 |