Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMC3
Entrez Gene ID
9126
Gene Name
structural maintenance of chromosomes 3
Gene Aliases
BAM, BMH, CDLS3, CSPG6, HCAP, SMC3L1
Chromosome Location
Chr.10: 110567691 - 110604634 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005445.3 NP_005436.1 17 - 18 1933 80
GenBank mRNA AF020043.1 - 17 - 18 1898 80
AF067163.1 - 5 - 6 581 80
AK289771.1 - 17 - 18 1929 80
AK315845.1 - 17 - 18 1932 80
BC047324.1 - 17 - 18 1828 80

View Full Product Details