Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK16
Entrez Gene ID
5127
Gene Name
cyclin dependent kinase 16
Gene Aliases
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
Chromosome Location
Chr.X: 47217881 - 47229997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170460.1 NP_001163931.1 13 - 14 1588 57
NM_006201.4 NP_006192.1 13 - 14 1723 57
NM_033018.3 NP_148978.2 13 - 14 1830 57
XM_011543920.2 XP_011542222.1 13 - 14 1600 57
XM_011543921.1 XP_011542223.1 14 - 15 1995 57
XM_011543922.1 XP_011542224.1 14 - 15 1983 57
XM_011543923.1 XP_011542225.1 13 - 14 1913 57
XM_011543924.1 XP_011542226.1 13 - 14 1723 57
XM_011543925.2 XP_011542227.1 14 - 15 1805 57
XM_011543926.1 XP_011542228.1 13 - 14 2766 57
XM_011543927.2 XP_011542229.1 14 - 15 1843 57
XM_011543928.1 XP_011542230.1 14 - 15 2198 57
XM_017029569.1 XP_016885058.1 14 - 15 1995 57
XM_017029570.1 XP_016885059.1 14 - 15 1805 57
XM_017029571.1 XP_016885060.1 13 - 14 2766 57
XM_017029572.1 XP_016885061.1 14 - 15 1843 57
XM_017029573.1 XP_016885062.1 14 - 15 2198 57
GenBank mRNA AB209465.1 - 10 - 11 1960 57
AK096881.1 - 13 - 14 1712 57
AK131486.1 - 9 - 10 2505 57
AK290145.1 - 13 - 14 1647 57
AK301832.1 - 13 - 14 1588 57
AK303849.1 - 14 - 15 1603 57
BC001048.2 - 13 - 14 1726 57
BC006190.2 - 13 - 14 1540 57
BC009852.2 - 11 - 12 986 57
BC015607.2 - 13 - 14 1693 57
BT006827.1 - 12 - 13 1288 57
X66363.1 - 13 - 14 1411 57

View Full Product Details