Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK16
Entrez Gene ID
5127
Gene Name
cyclin dependent kinase 16
Gene Aliases
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
Chromosome Location
Chr.X: 47217881 - 47229997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170460.1 NP_001163931.1 5 - 6 819 73
NM_006201.4 NP_006192.1 5 - 6 954 73
NM_033018.3 NP_148978.2 5 - 6 1061 73
XM_011543920.2 XP_011542222.1 5 - 6 831 73
XM_011543921.1 XP_011542223.1 6 - 7 1226 73
XM_011543922.1 XP_011542224.1 6 - 7 1214 73
XM_011543923.1 XP_011542225.1 5 - 6 1144 73
XM_011543924.1 XP_011542226.1 5 - 6 954 73
XM_011543925.2 XP_011542227.1 6 - 7 1036 73
XM_011543926.1 XP_011542228.1 5 - 6 1997 73
XM_011543927.2 XP_011542229.1 6 - 7 1074 73
XM_011543928.1 XP_011542230.1 6 - 7 1429 73
XM_017029569.1 XP_016885058.1 6 - 7 1226 73
XM_017029570.1 XP_016885059.1 6 - 7 1036 73
XM_017029571.1 XP_016885060.1 5 - 6 1997 73
XM_017029572.1 XP_016885061.1 6 - 7 1074 73
XM_017029573.1 XP_016885062.1 6 - 7 1429 73
GenBank mRNA AB209465.1 - 4 - 5 760 73
AK096881.1 - 5 - 6 943 73
AK131486.1 - 4 - 5 1224 73
AK290145.1 - 5 - 6 878 73
AK296880.1 - 5 - 6 855 73
AK301832.1 - 5 - 6 819 73
AK303849.1 - 6 - 7 813 73
BC001048.2 - 5 - 6 957 73
BC006190.2 - 5 - 6 771 73
BC009852.2 - 3 - 4 217 73
BC015607.2 - 5 - 6 924 73
BT006827.1 - 4 - 5 519 73
X66363.1 - 5 - 6 642 73

View Full Product Details