Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAMLD1
Entrez Gene ID
10046
Gene Name
mastermind like domain containing 1
Gene Aliases
CG1, CXorf6, F18, HYSP2
Chromosome Location
Chr.X: 150361422 - 150514178 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005491.4 NP_005482.2 2 - 3 235 65
XM_006724802.3 XP_006724865.1 4 - 5 583 65
XM_011531092.2 XP_011529394.1 4 - 5 583 65
XM_011531093.2 XP_011529395.1 5 - 6 713 65
XM_011531095.2 XP_011529397.1 4 - 5 583 65
GenBank mRNA AK302612.1 - 3 - 4 277 65
AK315217.1 - 2 - 3 108 65
DC320452.1 - 3 - 4 389 65
DR001800.1 - 3 - 4 488 65

View Full Product Details