The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177466.2 | NP_001170937.1 | 2 - 3 | 1904 | 73 |
NM_005491.4 | NP_005482.2 | 3 - 4 | 1979 | 73 | |
XM_006724802.3 | XP_006724865.1 | 5 - 6 | 2327 | 73 | |
XM_006724803.3 | XP_006724866.1 | 4 - 5 | 2252 | 73 | |
XM_017029188.1 | XP_016884677.1 | 5 - 6 | 2199 | 73 | |
GenBank mRNA | AK302612.1 | - | 5 - 6 | 1751 | 73 |
AK315217.1 | - | 3 - 4 | 1852 | 73 | |
BC136324.1 | - | 2 - 3 | 1889 | 73 | |
DA527328.1 | - | 1 - 2 | 58 | 73 |