The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291428.1 | NP_001278357.1 | 3 - 4 | 306 | 92 |
NM_001291429.1 | NP_001278358.1 | 2 - 3 | 254 | 92 | |
NM_004324.3 | NP_004315.1 | 3 - 4 | 306 | 92 | |
NM_138761.3 | NP_620116.1 | 3 - 4 | 306 | 92 | |
NM_138764.4 | NP_620119.2 | 3 - 4 | 306 | 92 | |
NR_027882.1 | 3 - 4 | 306 | 92 | ||
XM_017027077.1 | XP_016882566.1 | 3 - 4 | 315 | 92 | |
GenBank mRNA | AF007826.1 | - | 3 - 4 | 237 | 92 |
AF247393.1 | - | 3 - 4 | 237 | 92 | |
AJ417988.1 | - | 2 - 3 | 216 | 92 | |
AJ586910.1 | - | 2 - 3 | 172 | 92 | |
AK291076.1 | - | 3 - 4 | 286 | 92 | |
BC014175.2 | - | 3 - 4 | 289 | 92 | |
BE396495.1 | - | 3 - 4 | 306 | 92 | |
BM706954.1 | - | 1 - 2 | 119 | 92 | |
CN281003.1 | - | 3 - 4 | 259 | 92 | |
JX524562.1 | - | 2 - 3 | 186 | 92 | |
KF830846.1 | - | 2 - 3 | 186 | 92 | |
KF830847.1 | - | 2 - 3 | 186 | 92 | |
KF830848.1 | - | 2 - 3 | 186 | 92 | |
L22473.1 | - | 3 - 4 | 237 | 92 | |
L22474.1 | - | 3 - 4 | 237 | 92 |