The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002830.3 | NP_002821.1 | 20 - 21 | 2754 | 102 |
XM_011511560.2 | XP_011509862.1 | 20 - 21 | 2659 | 102 | |
XM_011511562.2 | XP_011509864.1 | 19 - 20 | 2593 | 102 | |
XM_011511563.2 | XP_011509865.1 | 10 - 11 | 2117 | 102 | |
XM_017004596.1 | XP_016860085.1 | 20 - 21 | 2680 | 102 | |
XM_017004597.1 | XP_016860086.1 | 20 - 21 | 2186 | 102 | |
XM_017004598.1 | XP_016860087.1 | 20 - 21 | 2659 | 102 | |
XM_017004599.1 | XP_016860088.1 | 20 - 21 | 2165 | 102 | |
XM_017004600.1 | XP_016860089.1 | 20 - 21 | 2144 | 102 | |
XM_017004601.1 | XP_016860090.1 | 19 - 20 | 2635 | 102 | |
XM_017004602.1 | XP_016860091.1 | 19 - 20 | 2614 | 102 | |
XM_017004603.1 | XP_016860092.1 | 19 - 20 | 2099 | 102 | |
XM_017004604.1 | XP_016860093.1 | 19 - 20 | 2569 | 102 | |
XM_017004605.1 | XP_016860094.1 | 19 - 20 | 2465 | 102 | |
XM_017004606.1 | XP_016860095.1 | 19 - 20 | 2590 | 102 | |
XM_017004607.1 | XP_016860096.1 | 17 - 18 | 2351 | 102 | |
XM_017004608.1 | XP_016860097.1 | 17 - 18 | 2308 | 102 | |
GenBank mRNA | AK314836.1 | - | 20 - 21 | 2566 | 102 |
BC010674.1 | - | 20 - 21 | 2410 | 102 | |
BX648614.1 | - | 14 - 15 | 2988 | 102 | |
M68941.1 | - | 20 - 21 | 2754 | 102 |