Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTHFD2L
Entrez Gene ID
441024
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like
Gene Aliases
-
Chromosome Location
Chr.4: 74123280 - 74303099 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144978.1 NP_001138450.1 4 - 5 637 74
XM_005265690.2 XP_005265747.1 5 - 6 680 74
XM_017008218.1 XP_016863707.1 4 - 5 1017 74
XM_017008219.1 XP_016863708.1 4 - 5 1017 74
XM_017008220.1 XP_016863709.1 4 - 5 1017 74
XM_017008221.1 XP_016863710.1 4 - 5 1017 74
XM_017008222.1 XP_016863711.1 4 - 5 1017 74
XM_017008223.1 XP_016863712.1 4 - 5 1017 74
XM_017008224.1 XP_016863713.1 4 - 5 2242 74
XM_017008225.1 XP_016863714.1 7 - 8 798 74
XM_017008226.1 XP_016863715.1 7 - 8 673 74
XM_017008227.1 XP_016863716.1 5 - 6 680 74
XM_017008228.1 XP_016863717.1 5 - 6 680 74
XM_017008229.1 XP_016863718.1 5 - 6 680 74
XM_017008230.1 XP_016863719.1 5 - 6 680 74
XM_017008231.1 XP_016863720.1 6 - 7 723 74
XM_017008232.1 XP_016863721.1 5 - 6 689 74
XM_017008233.1 XP_016863722.1 2 - 3 327 74
XM_017008234.1 XP_016863723.1 5 - 6 680 74
XM_017008235.1 XP_016863724.1 6 - 7 822 74
GenBank mRNA AK122799.1 - 2 - 3 1958 74
BC032771.1 - 5 - 6 596 74

View Full Product Details