Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPHLN1
Entrez Gene ID
51535
Gene Name
periphilin 1
Gene Aliases
CR, HSPC206, HSPC232
Chromosome Location
Chr.12: 42326126 - 42448621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143787.1 NP_001137259.1 1 - 2 84 78
NM_001143788.1 NP_001137260.1 1 - 2 84 78
NM_001143789.1 NP_001137261.1 1 - 2 84 78
NM_016488.6 NP_057572.5 1 - 2 84 78
NM_201439.1 NP_958847.1 1 - 2 84 78
XM_005268962.3 XP_005269019.1 1 - 2 103 78
XM_011538465.2 XP_011536767.1 1 - 2 62 78
XM_011538471.2 XP_011536773.1 1 - 2 103 78
XM_017019437.1 XP_016874926.1 1 - 2 62 78
XM_017019438.1 XP_016874927.1 1 - 2 62 78
XM_017019441.1 XP_016874930.1 1 - 2 62 78
XM_017019442.1 XP_016874931.1 1 - 2 62 78
XM_017019445.1 XP_016874934.1 1 - 2 103 78
XM_017019446.1 XP_016874935.1 1 - 2 102 78
XM_017019449.1 XP_016874938.1 1 - 2 103 78
XM_017019450.1 XP_016874939.1 1 - 2 103 78
GenBank mRNA AK055690.1 - 1 - 2 72 78
AK299951.1 - 1 - 2 66 78
AK303612.1 - 1 - 2 78
AK308022.1 - 1 - 2 58 78
AY039238.1 - 1 - 2 66 78
BC025306.1 - 1 - 2 51 78
CF242917.1 - 1 - 2 99 78
DC424443.1 - 1 - 2 69 78

View Full Product Details