Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PYGB
Entrez Gene ID
5834
Gene Name
phosphorylase, glycogen; brain
Gene Aliases
GPBB
Chromosome Location
Chr.20: 25248070 - 25298012 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002862.3 NP_002853.2 11 - 12 1511 98
GenBank mRNA AB209080.1 - 11 - 12 1469 98
AK299697.1 - 11 - 12 1435 98
AK309036.1 - 11 - 12 1509 98
BC017045.2 - 11 - 12 1486 98
BC030795.2 - 11 - 12 1487 98
J03544.1 - 11 - 12 1481 98
U47025.1 - 11 - 12 1436 98

View Full Product Details