Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PYGB
Entrez Gene ID
5834
Gene Name
phosphorylase, glycogen; brain
Gene Aliases
GPBB
Chromosome Location
Chr.20: 25248070 - 25298012 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002862.3 NP_002853.2 12 - 13 1627 77
GenBank mRNA AB209080.1 - 12 - 13 1585 77
AK299697.1 - 12 - 13 1551 77
AK309036.1 - 12 - 13 1625 77
BC017045.2 - 12 - 13 1602 77
BC030795.2 - 12 - 13 1603 77
J03544.1 - 12 - 13 1597 77
U47025.1 - 12 - 13 1552 77

View Full Product Details