Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGKB
Entrez Gene ID
1607
Gene Name
diacylglycerol kinase beta
Gene Aliases
DAGK2, DGK, DGK-BETA
Chromosome Location
Chr.7: 14145049 - 14974777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004080.2 NP_004071.1 7 - 8 780 89
NM_145695.2 NP_663733.1 7 - 8 780 89
XM_005249628.2 XP_005249685.1 8 - 9 1308 89
XM_005249629.2 XP_005249686.1 8 - 9 1308 89
XM_005249630.2 XP_005249687.1 7 - 8 821 89
XM_005249631.3 XP_005249688.1 7 - 8 1072 89
XM_011515153.2 XP_011513455.1 8 - 9 1146 89
XM_011515154.2 XP_011513456.1 8 - 9 862 89
XM_011515156.2 XP_011513458.1 7 - 8 821 89
XM_011515157.2 XP_011513459.1 8 - 9 861 89
XM_011515158.2 XP_011513460.1 7 - 8 997 89
XM_017011783.1 XP_016867272.1 8 - 9 1202 89
XM_017011784.1 XP_016867273.1 8 - 9 862 89
XM_017011785.1 XP_016867274.1 7 - 8 1093 89
XM_017011786.1 XP_016867275.1 7 - 8 1076 89
XM_017011787.1 XP_016867276.1 7 - 8 826 89
XM_017011788.1 XP_016867277.1 8 - 9 1145 89
XM_017011789.1 XP_016867278.1 8 - 9 862 89
XM_017011790.1 XP_016867279.1 8 - 9 1145 89
XM_017011791.1 XP_016867280.1 8 - 9 1144 89
XM_017011792.1 XP_016867281.1 8 - 9 1144 89
GenBank mRNA AB018261.1 - 4 - 5 409 89
AK295668.1 - 8 - 9 1114 89
AK304719.1 - 7 - 8 89
BC105005.1 - 7 - 8 660 89
BC143632.1 - 7 - 8 660 89
BC143633.1 - 7 - 8 660 89

View Full Product Details