Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H2AFY
Entrez Gene ID
9555
Gene Name
H2A histone family member Y
Gene Aliases
H2A.y, H2A/y, H2AF12M, MACROH2A1.1, mH2A1, macroH2A1.2
Chromosome Location
Chr.5: 135334381 - 135399887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138609.2 NP_613075.1 4 - 5 681 94
NM_138610.2 NP_613258.2 4 - 5 681 94
XM_005272134.3 XP_005272191.1 4 - 5 1059 94
XM_011543729.2 XP_011542031.1 4 - 5 598 94
XM_011543731.1 XP_011542033.1 4 - 5 1059 94
XM_011543733.2 XP_011542035.1 4 - 5 1059 94
XM_011543734.2 XP_011542036.1 4 - 5 1059 94
XM_017010068.1 XP_016865557.1 4 - 5 1059 94
GenBank mRNA AF041483.1 - 4 - 5 578 94
AF044286.1 - 3 - 4 473 94
AK023409.1 - 4 - 5 672 94
BC013331.1 - 4 - 5 607 94
BC095406.1 - 4 - 5 641 94

View Full Product Details