The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040158.1 | NP_001035248.1 | 5 - 6 | 795 | 87 |
NM_004893.2 | NP_004884.1 | 5 - 6 | 758 | 87 | |
NM_138610.2 | NP_613258.2 | 5 - 6 | 798 | 87 | |
XM_011543729.2 | XP_011542031.1 | 5 - 6 | 715 | 87 | |
XM_011543730.2 | XP_011542032.1 | 5 - 6 | 715 | 87 | |
XM_011543731.1 | XP_011542033.1 | 5 - 6 | 1176 | 87 | |
XM_011543732.1 | XP_011542034.1 | 5 - 6 | 1173 | 87 | |
XM_017010068.1 | XP_016865557.1 | 5 - 6 | 1176 | 87 | |
GenBank mRNA | AF041483.1 | - | 5 - 6 | 695 | 87 |
AF054174.1 | - | 5 - 6 | 758 | 87 | |
AY134746.1 | - | 4 - 5 | 571 | 87 | |
BC013331.1 | - | 5 - 6 | 724 | 87 | |
BC095406.1 | - | 5 - 6 | 758 | 87 |