The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020777.2 | NP_065828.2 | 14 - 15 | 1864 | 73 |
XM_005247987.4 | XP_005248044.2 | 14 - 15 | 1904 | 73 | |
XM_011513514.2 | XP_011511816.1 | 14 - 15 | 1904 | 73 | |
XM_011513515.2 | XP_011511817.1 | 14 - 15 | 1904 | 73 | |
XM_011513516.2 | XP_011511818.1 | 14 - 15 | 1904 | 73 | |
XM_017008481.1 | XP_016863970.1 | 14 - 15 | 1904 | 73 | |
GenBank mRNA | AB037750.1 | - | 11 - 12 | 1108 | 73 |
AK295629.1 | - | 14 - 15 | 1480 | 73 |