Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SORCS2
Entrez Gene ID
57537
Gene Name
sortilin related VPS10 domain containing receptor 2
Gene Aliases
-
Chromosome Location
Chr.4: 7192607 - 7742837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020777.2 NP_065828.2 11 - 12 1588 86
XM_005247987.4 XP_005248044.2 11 - 12 1628 86
XM_011513514.2 XP_011511816.1 11 - 12 1628 86
XM_011513515.2 XP_011511817.1 11 - 12 1628 86
XM_011513516.2 XP_011511818.1 11 - 12 1628 86
XM_017008481.1 XP_016863970.1 11 - 12 1628 86
GenBank mRNA AB037750.1 - 8 - 9 832 86
AK295629.1 - 11 - 12 1204 86

View Full Product Details