The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020777.2 | NP_065828.2 | 1 - 2 | 485 | 60 |
XM_005247987.4 | XP_005248044.2 | 1 - 2 | 525 | 60 | |
XM_011513514.2 | XP_011511816.1 | 1 - 2 | 525 | 60 | |
XM_011513515.2 | XP_011511817.1 | 1 - 2 | 525 | 60 | |
XM_011513516.2 | XP_011511818.1 | 1 - 2 | 525 | 60 | |
XM_017008481.1 | XP_016863970.1 | 1 - 2 | 525 | 60 | |
GenBank mRNA | AF286190.1 | - | 1 - 2 | 384 | 60 |
AI206155.1 | - | 1 - 2 | 387 | 60 |