The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316345.1 | NP_001303274.1 | 13 - 14 | 1523 | 109 |
NM_021111.2 | NP_066934.1 | 11 - 12 | 1387 | 109 | |
XM_017015207.1 | XP_016870696.1 | 12 - 13 | 1423 | 109 | |
XM_017015208.1 | XP_016870697.1 | 12 - 13 | 1497 | 109 | |
XM_017015209.1 | XP_016870698.1 | 12 - 13 | 1486 | 109 | |
XM_017015210.1 | XP_016870699.1 | 10 - 11 | 1238 | 109 | |
XM_017015211.1 | XP_016870700.1 | 11 - 12 | 1389 | 109 | |
GenBank mRNA | AK292653.1 | - | 11 - 12 | 1387 | 109 |
BC137093.1 | - | 11 - 12 | 1348 | 109 | |
BX648668.1 | - | 9 - 10 | 1265 | 109 | |
D50406.1 | - | 11 - 12 | 1410 | 109 |