The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316345.1 | NP_001303274.1 | 5 - 6 | 388 | 79 |
NM_001316346.1 | NP_001303275.1 | 4 - 5 | 354 | 79 | |
NM_001316347.1 | NP_001303276.1 | 4 - 5 | 354 | 79 | |
NM_001316348.1 | NP_001303277.1 | 4 - 5 | 354 | 79 | |
NM_021111.2 | NP_066934.1 | 4 - 5 | 354 | 79 | |
XM_017015207.1 | XP_016870696.1 | 5 - 6 | 390 | 79 | |
XM_017015208.1 | XP_016870697.1 | 4 - 5 | 362 | 79 | |
XM_017015211.1 | XP_016870700.1 | 4 - 5 | 356 | 79 | |
GenBank mRNA | AK315073.1 | - | 4 - 5 | 340 | 79 |
BC060806.1 | - | 4 - 5 | 328 | 79 | |
BC137093.1 | - | 4 - 5 | 315 | 79 | |
BX393623.2 | - | 4 - 5 | 342 | 79 | |
D50406.1 | - | 4 - 5 | 377 | 79 | |
JQ756320.1 | - | 4 - 5 | 268 | 79 |