The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177310.1 | NP_001170781.1 | 21 - 22 | 2975 | 88 |
NM_001177311.1 | NP_001170782.1 | 21 - 22 | 3002 | 88 | |
NM_017662.4 | NP_060132.3 | 21 - 22 | 3156 | 88 | |
XM_011518244.1 | XP_011516546.1 | 21 - 22 | 3156 | 88 | |
XM_011518245.1 | XP_011516547.1 | 20 - 21 | 3063 | 88 | |
XM_011518246.1 | XP_011516548.1 | 21 - 22 | 3156 | 88 | |
XM_011518247.1 | XP_011516549.1 | 20 - 21 | 3027 | 88 | |
XM_011518248.1 | XP_011516550.1 | 20 - 21 | 3015 | 88 | |
XM_011518249.1 | XP_011516551.1 | 19 - 20 | 2922 | 88 | |
XM_011518250.1 | XP_011516552.1 | 19 - 20 | 2880 | 88 | |
XM_011518251.2 | XP_011516553.1 | 16 - 17 | 2407 | 88 | |
XM_011518252.2 | XP_011516554.1 | 21 - 22 | 3156 | 88 | |
XM_011518255.2 | XP_011516557.1 | 21 - 22 | 3156 | 88 | |
XM_017014287.1 | XP_016869776.1 | 18 - 19 | 2793 | 88 | |
XM_017014288.1 | XP_016869777.1 | 17 - 18 | 2646 | 88 | |
XM_017014289.1 | XP_016869778.1 | 21 - 22 | 3156 | 88 | |
GenBank mRNA | AF350881.2 | - | 21 - 22 | 3156 | 88 |
AF448232.1 | - | 21 - 22 | 2918 | 88 | |
AK026281.1 | - | 15 - 16 | 2223 | 88 | |
AY333282.1 | - | 21 - 22 | 2981 | 88 | |
AY333283.1 | - | 21 - 22 | 2975 | 88 | |
AY333284.1 | - | 21 - 22 | 3002 | 88 | |
AY333285.1 | - | 21 - 22 | 2981 | 88 |