The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177310.1 | NP_001170781.1 | 31 - 32 | 5142 | 79 |
NM_001177311.1 | NP_001170782.1 | 31 - 32 | 5169 | 79 | |
NM_017662.4 | NP_060132.3 | 31 - 32 | 5323 | 79 | |
XM_011518244.1 | XP_011516546.1 | 31 - 32 | 5323 | 79 | |
XM_011518245.1 | XP_011516547.1 | 30 - 31 | 5230 | 79 | |
XM_011518246.1 | XP_011516548.1 | 31 - 32 | 5323 | 79 | |
XM_011518247.1 | XP_011516549.1 | 30 - 31 | 5194 | 79 | |
XM_011518248.1 | XP_011516550.1 | 30 - 31 | 5182 | 79 | |
XM_011518249.1 | XP_011516551.1 | 29 - 30 | 5089 | 79 | |
XM_011518250.1 | XP_011516552.1 | 29 - 30 | 5047 | 79 | |
XM_011518251.2 | XP_011516553.1 | 26 - 27 | 4574 | 79 | |
XM_017014287.1 | XP_016869776.1 | 28 - 29 | 4960 | 79 | |
XM_017014288.1 | XP_016869777.1 | 27 - 28 | 4813 | 79 | |
GenBank mRNA | AF350881.2 | - | 31 - 32 | 5323 | 79 |
AF448232.1 | - | 31 - 32 | 5085 | 79 | |
AK000094.1 | - | 8 - 9 | 2049 | 79 | |
AK225207.1 | - | 8 - 9 | 2049 | 79 | |
AY333282.1 | - | 31 - 32 | 5148 | 79 | |
AY333283.1 | - | 31 - 32 | 5142 | 79 | |
AY333284.1 | - | 31 - 32 | 5169 | 79 | |
AY333285.1 | - | 31 - 32 | 5148 | 79 | |
AY333286.1 | - | 18 - 19 | 2001 | 79 | |
AY333287.1 | - | 15 - 16 | 1647 | 79 |