The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177310.1 | NP_001170781.1 | 34 - 35 | 5549 | 144 |
NM_001177311.1 | NP_001170782.1 | 34 - 35 | 5576 | 144 | |
NM_017662.4 | NP_060132.3 | 34 - 35 | 5730 | 144 | |
XM_011518244.1 | XP_011516546.1 | 34 - 35 | 5727 | 144 | |
XM_011518245.1 | XP_011516547.1 | 33 - 34 | 5637 | 144 | |
XM_011518246.1 | XP_011516548.1 | 35 - 36 | 5607 | 144 | |
XM_011518247.1 | XP_011516549.1 | 33 - 34 | 5601 | 144 | |
XM_011518248.1 | XP_011516550.1 | 33 - 34 | 5589 | 144 | |
XM_011518249.1 | XP_011516551.1 | 32 - 33 | 5496 | 144 | |
XM_011518250.1 | XP_011516552.1 | 32 - 33 | 5454 | 144 | |
XM_011518251.2 | XP_011516553.1 | 29 - 30 | 4981 | 144 | |
XM_017014287.1 | XP_016869776.1 | 31 - 32 | 5367 | 144 | |
XM_017014288.1 | XP_016869777.1 | 30 - 31 | 5220 | 144 | |
GenBank mRNA | AF350881.2 | - | 34 - 35 | 5730 | 144 |
AF448232.1 | - | 34 - 35 | 5492 | 144 | |
AK000094.1 | - | 11 - 12 | 2456 | 144 | |
AK225207.1 | - | 11 - 12 | 2456 | 144 | |
AY333282.1 | - | 34 - 35 | 5555 | 144 | |
AY333283.1 | - | 34 - 35 | 5549 | 144 | |
AY333284.1 | - | 34 - 35 | 5576 | 144 | |
AY333285.1 | - | 34 - 35 | 5555 | 144 | |
AY333286.1 | - | 21 - 22 | 2408 | 144 | |
AY333287.1 | - | 18 - 19 | 2054 | 144 | |
AY333288.1 | - | 8 - 9 | 1196 | 144 |