The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037872.2 | NP_001032961.1 | 5 - 6 | 755 | 92 |
NM_001321454.1 | NP_001308383.1 | 5 - 6 | 755 | 92 | |
NM_001321455.1 | NP_001308384.1 | 6 - 7 | 912 | 92 | |
NM_001321458.1 | NP_001308387.1 | 5 - 6 | 755 | 92 | |
NM_001321459.1 | NP_001308388.1 | 5 - 6 | 755 | 92 | |
NM_001321460.1 | NP_001308389.1 | 5 - 6 | 755 | 92 | |
NM_016316.3 | NP_057400.1 | 5 - 6 | 755 | 92 | |
NR_135649.1 | 5 - 6 | 755 | 92 | ||
NR_135650.1 | 6 - 7 | 912 | 92 | ||
NR_135651.1 | 5 - 6 | 755 | 92 | ||
NR_135652.1 | 5 - 6 | 755 | 92 | ||
NR_135653.1 | 5 - 6 | 755 | 92 | ||
XM_017004309.1 | XP_016859798.1 | 5 - 6 | 1168 | 92 | |
XM_017004310.1 | XP_016859799.1 | 5 - 6 | 534 | 92 | |
XM_017004311.1 | XP_016859800.1 | 5 - 6 | 534 | 92 | |
XM_017004312.1 | XP_016859801.1 | 5 - 6 | 750 | 92 | |
XM_017004313.1 | XP_016859802.1 | 5 - 6 | 2484 | 92 | |
GenBank mRNA | AB047646.1 | - | 5 - 6 | 680 | 92 |
AF151538.1 | - | 5 - 6 | 708 | 92 | |
AF206019.1 | - | 5 - 6 | 717 | 92 | |
AF357886.1 | - | 5 - 6 | 673 | 92 | |
AK002087.1 | - | 3 - 4 | 617 | 92 | |
AK307719.1 | - | 5 - 6 | 666 | 92 | |
BC037734.1 | - | 6 - 7 | 893 | 92 | |
BC130411.1 | - | 4 - 5 | 526 | 92 | |
DA852908.1 | - | 2 - 3 | 253 | 92 |