Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPHLN1
Entrez Gene ID
51535
Gene Name
periphilin 1
Gene Aliases
CR, HSPC206, HSPC232
Chromosome Location
Chr.12: 42326126 - 42448621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143787.1 NP_001137259.1 9 - 10 961 116
NM_016488.6 NP_057572.5 10 - 11 1126 116
XM_005268962.3 XP_005269019.1 8 - 9 923 116
XM_011538459.2 XP_011536761.1 11 - 12 1147 116
XM_011538465.2 XP_011536767.1 9 - 10 1047 116
XM_011538467.2 XP_011536769.1 10 - 11 986 116
XM_011538470.2 XP_011536772.1 9 - 10 947 116
XM_011538471.2 XP_011536773.1 8 - 9 923 116
XM_017019434.1 XP_016874923.1 11 - 12 1147 116
XM_017019435.1 XP_016874924.1 11 - 12 1146 116
XM_017019436.1 XP_016874925.1 11 - 12 1147 116
XM_017019437.1 XP_016874926.1 10 - 11 1104 116
XM_017019438.1 XP_016874927.1 10 - 11 1104 116
XM_017019439.1 XP_016874928.1 11 - 12 1147 116
XM_017019440.1 XP_016874929.1 10 - 11 1090 116
XM_017019441.1 XP_016874930.1 9 - 10 1047 116
XM_017019442.1 XP_016874931.1 9 - 10 1047 116
XM_017019443.1 XP_016874932.1 10 - 11 1090 116
XM_017019444.1 XP_016874933.1 10 - 11 986 116
XM_017019445.1 XP_016874934.1 9 - 10 980 116
XM_017019446.1 XP_016874935.1 9 - 10 979 116
XM_017019447.1 XP_016874936.1 9 - 10 947 116
XM_017019448.1 XP_016874937.1 9 - 10 947 116
XM_017019449.1 XP_016874938.1 8 - 9 923 116
XM_017019450.1 XP_016874939.1 8 - 9 923 116
XM_017019451.1 XP_016874940.1 9 - 10 947 116
GenBank mRNA AK299951.1 - 8 - 9 886 116
AK303612.1 - 9 - 10 961 116
AK303680.1 - 7 - 8 842 116
AY039238.1 - 10 - 11 1108 116
BC025306.1 - 9 - 10 928 116

View Full Product Details