Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO2
Entrez Gene ID
57101
Gene Name
anoctamin 2
Gene Aliases
C12orf3, TMEM16B
Chromosome Location
Chr.12: 5560241 - 5948790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278596.1 NP_001265525.1 18 - 19 1875 64
NM_001278597.1 NP_001265526.1 18 - 19 1893 64
XM_006718988.3 XP_006719051.1 16 - 17 2150 64
XM_011520975.2 XP_011519277.1 16 - 17 4436 64
XM_011520978.2 XP_011519280.1 15 - 16 1748 64
XM_017019672.1 XP_016875161.1 16 - 17 2150 64
GenBank mRNA AJ272204.1 - 18 - 19 1893 64
AK024010.1 - 1 - 2 157 64
FJ384095.1 - 18 - 19 1849 64

View Full Product Details