The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278596.1 | NP_001265525.1 | 19 - 20 | 1987 | 86 |
NM_001278597.1 | NP_001265526.1 | 19 - 20 | 2005 | 86 | |
XM_006718988.3 | XP_006719051.1 | 17 - 18 | 2262 | 86 | |
XM_011520975.2 | XP_011519277.1 | 17 - 18 | 4548 | 86 | |
XM_011520978.2 | XP_011519280.1 | 16 - 17 | 1860 | 86 | |
XM_017019672.1 | XP_016875161.1 | 17 - 18 | 2262 | 86 | |
GenBank mRNA | AJ272204.1 | - | 19 - 20 | 2005 | 86 |
AK024010.1 | - | 2 - 3 | 269 | 86 | |
FJ384095.1 | - | 19 - 20 | 1961 | 86 |