Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO2
Entrez Gene ID
57101
Gene Name
anoctamin 2
Gene Aliases
C12orf3, TMEM16B
Chromosome Location
Chr.12: 5560241 - 5948790 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278596.1 NP_001265525.1 3 - 4 580 65
NM_001278597.1 NP_001265526.1 3 - 4 598 65
XM_006718988.3 XP_006719051.1 3 - 4 870 65
XM_011520975.2 XP_011519277.1 3 - 4 3156 65
XM_017019672.1 XP_016875161.1 3 - 4 870 65
GenBank mRNA AB593159.1 - 3 - 4 580 65
AJ272204.1 - 3 - 4 598 65
FJ384095.1 - 3 - 4 554 65

View Full Product Details