The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278596.1 | NP_001265525.1 | 6 - 7 | 830 | 109 |
NM_001278597.1 | NP_001265526.1 | 6 - 7 | 848 | 109 | |
XM_006718988.3 | XP_006719051.1 | 5 - 6 | 1117 | 109 | |
XM_011520975.2 | XP_011519277.1 | 5 - 6 | 3403 | 109 | |
XM_011520978.2 | XP_011519280.1 | 4 - 5 | 715 | 109 | |
XM_017019672.1 | XP_016875161.1 | 5 - 6 | 1117 | 109 | |
GenBank mRNA | AB593159.1 | - | 5 - 6 | 827 | 109 |
AJ272204.1 | - | 6 - 7 | 848 | 109 | |
DB071682.1 | - | 3 - 4 | 453 | 109 | |
FJ384095.1 | - | 6 - 7 | 804 | 109 |