Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPIFB6
Entrez Gene ID
128859
Gene Name
BPI fold containing family B member 6
Gene Aliases
BPIL3, LPLUNC6
Chromosome Location
Chr.20: 33031648 - 33044108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174897.2 NP_777557.1 2 - 3 194 65
XM_017027663.1 XP_016883152.1 2 - 3 242 65
GenBank mRNA AF465767.1 - 2 - 3 194 65

View Full Product Details