The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099.4 | NP_001090.2 | 3 - 4 | 394 | 108 |
NM_001134194.1 | NP_001127666.1 | 3 - 4 | 394 | 108 | |
NM_001292037.1 | NP_001278966.1 | 3 - 4 | 394 | 108 | |
XM_011512946.1 | XP_011511248.1 | 3 - 4 | 358 | 108 | |
XM_011512947.2 | XP_011511249.1 | 3 - 4 | 358 | 108 | |
GenBank mRNA | AB102888.1 | - | 3 - 4 | 304 | 108 |
AK300540.1 | - | 3 - 4 | 355 | 108 | |
BC007460.1 | - | 3 - 4 | 354 | 108 | |
BC008493.1 | - | 3 - 4 | 351 | 108 | |
BC016344.1 | - | 3 - 4 | 358 | 108 | |
BC017877.1 | - | 3 - 4 | 344 | 108 | |
DC383453.1 | - | 3 - 4 | 394 | 108 | |
M24902.1 | - | 3 - 4 | 327 | 108 | |
M34840.1 | - | 3 - 4 | 318 | 108 | |
X52174.1 | - | 3 - 4 | 327 | 108 | |
X53605.1 | - | 3 - 4 | 340 | 108 |