Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WWC2
Entrez Gene ID
80014
Gene Name
WW and C2 domain containing 2
Gene Aliases
BOMB
Chromosome Location
Chr.4: 183098677 - 183320777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024949.5 NP_079225.5 16 - 17 2748 70
XM_011532269.2 XP_011530571.1 16 - 17 3381 70
GenBank mRNA AJ566366.1 - 3 - 4 430 70
AK074260.1 - 3 - 4 460 70
AK126057.1 - 17 - 18 2642 70
AK127061.1 - 9 - 10 1660 70
BC053873.1 - 7 - 8 1177 70
BC107683.1 - 7 - 8 1345 70
BX647378.1 - 14 - 15 2339 70
BX647704.1 - 4 - 5 475 70

View Full Product Details