The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024949.5 | NP_079225.5 | 5 - 6 | 786 | 114 |
XM_011532269.2 | XP_011530571.1 | 5 - 6 | 1419 | 114 | |
GenBank mRNA | AK126057.1 | - | 4 - 5 | 451 | 114 |
BM046889.1 | - | 3 - 4 | 222 | 114 |