The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024949.5 | NP_079225.5 | 6 - 7 | 915 | 75 |
XM_011532269.2 | XP_011530571.1 | 6 - 7 | 1548 | 75 | |
GenBank mRNA | AK126057.1 | - | 5 - 6 | 580 | 75 |
BM046889.1 | - | 4 - 5 | 351 | 75 | |
BX647378.1 | - | 4 - 5 | 506 | 75 |