Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
Chromosome Location
Chr.11: 119044185 - 119057246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130991.2 NP_001124463.1 3 - 4 290 63
NM_006389.4 NP_006380.1 3 - 4 423 63
XM_005271392.3 XP_005271449.1 3 - 4 284 63
XM_005271393.3 XP_005271450.1 3 - 4 264 63
XM_005271394.3 XP_005271451.1 3 - 4 331 63
XM_017017095.1 XP_016872584.1 3 - 4 319 63
XM_017017096.1 XP_016872585.1 3 - 4 340 63
XM_017017097.1 XP_016872586.1 3 - 4 263 63
GenBank mRNA AK222756.1 - 3 - 4 316 63
AK294905.1 - 4 - 5 451 63
AK297260.1 - 3 - 4 249 63
AK299648.1 - 3 - 4 246 63
AK314178.1 - 3 - 4 314 63
BC004560.2 - 3 - 4 238 63
BC017726.1 - 3 - 4 241 63
BC072436.1 - 3 - 4 289 63
BP236985.1 - 3 - 4 258 63
BP358970.1 - 3 - 4 382 63
DC364982.1 - 3 - 4 382 63
DQ350134.1 - 3 - 4 311 63
DQ372932.1 - 3 - 4 311 63
U65785.1 - 3 - 4 291 63

View Full Product Details