The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130991.2 | NP_001124463.1 | 6 - 7 | 596 | 152 |
NM_006389.4 | NP_006380.1 | 6 - 7 | 729 | 152 | |
XM_005271392.3 | XP_005271449.1 | 6 - 7 | 590 | 152 | |
XM_005271393.3 | XP_005271450.1 | 6 - 7 | 570 | 152 | |
XM_005271394.3 | XP_005271451.1 | 6 - 7 | 637 | 152 | |
XM_017017095.1 | XP_016872584.1 | 6 - 7 | 625 | 152 | |
XM_017017096.1 | XP_016872585.1 | 6 - 7 | 646 | 152 | |
XM_017017097.1 | XP_016872586.1 | 6 - 7 | 569 | 152 | |
GenBank mRNA | AK127324.1 | - | 5 - 6 | 598 | 152 |
AK222756.1 | - | 6 - 7 | 622 | 152 | |
AK294905.1 | - | 7 - 8 | 757 | 152 | |
AK301514.1 | - | 4 - 5 | 288 | 152 | |
AK304264.1 | - | 5 - 6 | 528 | 152 | |
AK307624.1 | - | 4 - 5 | 211 | 152 | |
AK314178.1 | - | 6 - 7 | 620 | 152 | |
BC017726.1 | - | 6 - 7 | 547 | 152 | |
BC072436.1 | - | 6 - 7 | 595 | 152 | |
DQ350134.1 | - | 6 - 7 | 617 | 152 | |
DQ372932.1 | - | 6 - 7 | 617 | 152 | |
U65785.1 | - | 6 - 7 | 597 | 152 |