Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
Chromosome Location
Chr.11: 119044185 - 119057246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130991.2 NP_001124463.1 7 - 8 781 80
NM_006389.4 NP_006380.1 7 - 8 914 80
XM_005271392.3 XP_005271449.1 7 - 8 775 80
XM_005271393.3 XP_005271450.1 7 - 8 755 80
XM_005271394.3 XP_005271451.1 7 - 8 822 80
XM_017017095.1 XP_016872584.1 7 - 8 810 80
XM_017017096.1 XP_016872585.1 7 - 8 831 80
XM_017017097.1 XP_016872586.1 7 - 8 754 80
GenBank mRNA AK127324.1 - 6 - 7 783 80
AK222756.1 - 7 - 8 807 80
AK294905.1 - 8 - 9 942 80
AK301514.1 - 5 - 6 473 80
AK304264.1 - 6 - 7 713 80
AK307624.1 - 5 - 6 396 80
AK314178.1 - 7 - 8 805 80
BC017726.1 - 7 - 8 732 80
BC072436.1 - 7 - 8 780 80
DQ350134.1 - 7 - 8 802 80
DQ372932.1 - 7 - 8 802 80
U65785.1 - 7 - 8 782 80

View Full Product Details