The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020733.1 | NP_065784.1 | 10 - 11 | 3420 | 83 |
XM_005247666.1 | XP_005247723.1 | 11 - 12 | 3715 | 83 | |
GenBank mRNA | AB033063.2 | - | 9 - 10 | 3015 | 83 |