Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMARCC1
Entrez Gene ID
6599
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1
Gene Aliases
BAF155, CRACC1, Rsc8, SRG3, SWI3
Chromosome Location
Chr.3: 47585888 - 47781915 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003074.3 NP_003065.3 16 - 17 1692 67
XM_011534034.1 XP_011532336.1 16 - 17 1423 67
GenBank mRNA AK302412.1 - 2 - 3 136 67
AK302541.1 - 16 - 17 1467 67
BC021862.1 - 16 - 17 1629 67
BC032564.1 - 16 - 17 1676 67
BC039843.1 - 16 - 17 1596 67
BC040242.1 - 1 - 2 124 67
BC050564.1 - 16 - 17 1621 67
BC065253.1 - 16 - 17 1661 67
BC113465.1 - 16 - 17 1640 67
BC117213.1 - 16 - 17 1640 67

View Full Product Details